missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Folpet, MilliporeSigma™ Supelco™
Description
Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 133-07-3 |
| Physical Form | Neat |
| Linear Formula | C9H4 Cl3 NO2 S |
| MDL Number | MFCD00047303 |
| Synonym | 2-(Trichloromethylsulfanyl)isoindole-1,3-dione |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 296.56 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH