missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Flupyrsulfuron-methyl sodium, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 144740-54-5 |
| Physical Form | Neat |
| Linear Formula | C15H13F3N5 NaO7 S |
| Synonym | Methyl 2-[(4,6-dimethoxypyrimidin-2-ylcarbamoyl)sulfamoyl]-6-(trifluoromethyl)nicotinate monosodium salt monosodium salt |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 487.34 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH