missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Fluorodifen, MilliporeSigma™ Supelco™

Catalog No. 111020633
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020633 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020633 Supplier Merck Emd Millipore Supplier No. 45506250MG
Only null left
Agriculture

Specifications

Product Line PESTANAL
CAS 15457-05-3
Physical Form Neat
Linear Formula C13H7F3N2O5
MDL Number MFCD00055532
Type Analytical Standard
Quantity 250 mg
Formula Weight 328.20
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH