missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Fluopyram, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 658066-35-4 |
| Physical Form | Neat |
| Linear Formula | C16H11 ClF6N2 O |
| MDL Number | MFCD17166960 |
| Synonym | N-[2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]ethyl]-2-(trifluoromethyl)benzamide; N-{2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]ethyl}-alpha,alpha,alpha-trifluoro-o-toluamide |
| Type | Analytical Standard |
| Quantity | 50 mg |
| Formula Weight | 396.71 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH