missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Fluopicolide, MilliporeSigma™ Supelco™

Catalog No. 111018554
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018554 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018554 Supplier Merck Emd Millipore Supplier No. 41132100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Agriculture

Specifications

Product Line PESTANAL
CAS 239110-15-7
Physical Form Neat
Linear Formula C14H8 Cl3F3N2 O
MDL Number MFCD09751509
Synonym 2,6-Dichloro-N-[3-chloro-5-(trifluoromethyl)-2-pyridylmethyl]benzamide
Type Analytical Standard
Quantity 100 mg
Formula Weight 383.58
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH