missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Flucythrinate, Mixture of stereo isomers, MilliporeSigma™ Supelco™

Catalog No. 111018926
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018926 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018926 Supplier Merck Emd Millipore Supplier No. 33496100MGR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Flucythrinate belongs to the class of synthetic pyrethroids, primarily used to control Heliothis spp. in cotton and other insect pests in agricultural crops.

Agriculture

Specifications

Product Line PESTANAL
CAS 70124-77-5
Physical Form Neat
Linear Formula C26H23F2 NO4
MDL Number MFCD00137383
Synonym alpha-Cyano-3-phenoxybenzyl 4-difluoromethoxy-alpha-isopropylphenylacetate
Type Analytical Standard
Quantity 100 mg
Formula Weight 451.46
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH