missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Fluazinam, MilliporeSigma™ Supelco™

Catalog No. 111018974
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018974 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018974 Supplier Merck Emd Millipore Supplier No. 34095100MG
Only null left

Fluazinam is a 2,6-dinitroaniline protectant fungicide, which belongs to the pyridinamine group. It can mostly find applications in agriculture.

Agriculture

Specifications

Product Line PESTANAL
CAS 79622-59-6
Physical Form Neat
Linear Formula C13H4 Cl2F6N4O4
MDL Number MFCD00214168
Synonym 3-Chloro-N-(3-chloro-5-trifluoromethyl-2-pyridyl)-alpha,alpha,alpha-trifluoro-2,6-dinitro-p-toluidine; 3-Chloro-N-(3-chloro-2,6-dinitro-4-trifluoromethylphenyl)-5-trifluoromethyl-2-pyridylamine
Type Analytical Standard
Quantity 100 mg
Formula Weight 465.09
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH