missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Flazasulfuron, MilliporeSigma™ Supelco™

Catalog No. 111018360
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018360 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018360 Supplier Merck Emd Millipore Supplier No. 3405250MGR
Only null left

Flazasulfuron may generally be used as a sulfonylurea herbicide, which is having broad spectrum applications in weed control management. Its mode of action involves the inhibition of branched-chain amino acids, via binding irreversibly to to acetolactate synthase (ALS).

Agriculture

Specifications

Product Line PESTANAL
CAS 104040-78-0
Physical Form Neat
Linear Formula C13H12F3N5O5 S
MDL Number MFCD00274594
Synonym N-(4,6-Dimethoxy-2-pyrimidinylaminocarbonyl)-3-(trifluoromethyl)-2-pyridinesulfonamide
Type Analytical Standard
Quantity 50 mg
Formula Weight 407.33
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH