missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Fipronil sulfide, MilliporeSigma™ Supelco™
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 120067-83-6 |
| Linear Formula | C12H4 Cl2F6N4 S |
| MDL Number | MFCD08273815 |
| Synonym | 5-Amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-trifluoromethylsulfanyl-1 H-pyrazole-3-carbonitrile; 5-Amino-3-cyano-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-(trifluoromethylthio)pyrazole |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 421.15 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH