missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Fipronil-desulfinyl, ≥94.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018170
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018170 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018170 Supplier Merck Emd Millipore Supplier No. 4186525MG

Find more information here fipronil

Agriculture

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
Product Line PESTANAL
CAS 205650-65-3
Physical Form Neat
Linear Formula C12H4 Cl2F6N4
MDL Number MFCD08273842
Melting Point 189°C to 190°C
Synonym 5-Amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-(trifluoromethyl)-1 H-pyrazole-3-carbonitrile
Type Analytical Standard
Quantity 25 mg
Formula Weight 389.08
Percent Purity ≥94.0% (HPLC)
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH