missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Fenthion oxon, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018290
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018290 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018290 Supplier Merck Emd Millipore Supplier No. 892575MG
Only null left

Specifications

CAS 6552-12-1
Product Line PESTANAL
Linear Formula C10H15O4 PS
MDL Number MFCD00155408
Synonym O,O′-Dimethyl O′′-[3-methyl-4-(methylthio)phenyl] phosphate; Dimethyl 4-(methylthio)-m-tolyl phosphate; Fenoxon; Fenthoxon
Type Analytical Standard
Quantity 5 mg
Formula Weight 262.26
Percent Purity ≥98% (HPLC)
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH