missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Fensulfothion oxon, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 6552-21-2 |
| Linear Formula | C11H17O5 PS |
| MDL Number | MFCD01705553 |
| Synonym | Diethyl 4-(methylsulfinyl)phenyl phosphate; Fensulfothion PO-sulfoxide; NSC 133045 |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 292.29 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH