missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Fensulfothion, MilliporeSigma™ Supelco™

Catalog No. 111020220
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020220 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020220 Supplier Merck Emd Millipore Supplier No. 45307100MG
Only null left
Agriculture and Environmental

Specifications

Product Line PESTANAL
CAS 115-90-2
Physical Form Neat
Linear Formula C11H17O4 PS2
MDL Number MFCD00055487
Synonym Diethyl 4-(methylthio)phenyl phosphate
Type Analytical Standard
Quantity 100 mg
Formula Weight 308.35
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH