missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Fenpropathrin, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 39515-41-8 |
| Physical Form | Neat |
| Linear Formula | C22H23 NO3 |
| MDL Number | MFCD00144305 |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 349.42 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH