missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Fenpiclonil, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 74738-17-3 |
| Physical Form | Neat |
| Linear Formula | C11H6 Cl2N2 |
| MDL Number | MFCD00144304 |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 237.08 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH