missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Fenoxanil, MilliporeSigma™ Supelco™

Catalog No. 111012651
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012651 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012651 Supplier Merck Emd Millipore Supplier No. 33872100MGR
Only null left

Fenoxanil is a systemic fungicide used for the control of rice blast.

Agriculture

Specifications

Product Line PESTANAL
CAS 115852-48-7
Physical Form Neat
Linear Formula C15H18 Cl2N2O2
MDL Number MFCD06798290
Synonym N-(1-Cyano-1,2-dimethylpropyl)-2-(2,4-dichlorophenoxy)propionamide
Type Analytical Standard
Quantity 100 mg
Formula Weight 329.22
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH