missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Ethirimol, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 23947-60-6 |
| Physical Form | Neat |
| Linear Formula | C11H19N3 O |
| MDL Number | MFCD00055519 |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 209.29 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH