missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Ethion, MilliporeSigma™ Supelco™

Catalog No. 111020534
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020534 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020534 Supplier Merck Emd Millipore Supplier No. 45477250MG
Agriculture

Specifications

Product Line PESTANAL
CAS 563-12-2
Physical Form Neat
Linear Formula C9H22O4P2S4
MDL Number MFCD00036232
Type Analytical Standard
Quantity 250 mg
Formula Weight 384.48
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH