missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Etaconazol, MilliporeSigma™ Supelco™

Catalog No. 111015324
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015324 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015324 Supplier Merck Emd Millipore Supplier No. 45471250MG
Only null left
Agriculture

Specifications

Product Line PESTANAL
CAS 71245-23-3
Physical Form Neat
Linear Formula C14H15 Cl2N3O2
MDL Number MFCD00078700
Type Analytical Standard
Quantity 250 mg
Formula Weight 328.19
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH