missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ EPA 8151 Herbicide Acid Mix in Acetone (Varied), Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111017211
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017211 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017211 Supplier Merck Emd Millipore Supplier No. 48996U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is now available as a certified reference material. Please reference CRM48996.

Specifications

Packaging Ampule
Product Line PESTANAL
Type Organic Pollutant Standard
Quantity 1 mL
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) Acifluorfen 100 μg/mL; Bentazon 100 μg/mL; Chloramben 100 μg/mL; 2,4-D 100 μg/mL; 2,4-DB acid 100 μg/mL; Dalapon 100 μg/mL; Tetrachloroterephthalic acid; Dicamba 100 μg/mL; 3,5-Dichlorobenzoic acid 100 μg/mL; Dichloroprop 100 μg/mL; Dinoseb 100 μg/mL; MCPA 10000 μg/mL; MCPP 10000 μg/mL; 4-Nitrophenol 100 μg/mL; Pentachlorophenol 100 μg/mL; Picloram 100 μg/mL; 2,4,5-T 100 μg/mL; 2,4,5-TP 100 μg/mL

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH