missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Endrin ketone, MilliporeSigma™ Supelco™

Catalog No. 111019164
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019164 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019164 Supplier Merck Emd Millipore Supplier No. 442579

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Endrin ketone is classified under the group of organochlorine pesticides (OCPs) used in agriculture for the control of pests and diseases.

Agriculture

Specifications

Packaging Ampule
Product Line PESTANAL
CAS 53494-70-5
Physical Form Neat
Linear Formula C12H8 Cl6 O
Type Analytical Standard
Quantity 25 mg
Formula Weight 380.91
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH