missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Endosulfan ether, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 3369-52-6 |
| Physical Form | Neat |
| Linear Formula | C9H6 Cl6 O |
| MDL Number | MFCD00168052 |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 342.86 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH