missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Edifenphos, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 17109-49-8 |
| Physical Form | Neat |
| Linear Formula | C14H15O2 PS2 |
| MDL Number | MFCD00078673 |
| Synonym | O-Ethyl S,S-diphenyl phosphorodithioate |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 310.37 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH