missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ (E)-Metominostrobin, MilliporeSigma™ Supelco™

Catalog No. 111012956
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012956 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012956 Supplier Merck Emd Millipore Supplier No. 3423010MG
Only null left

Metominostrobin is a systemic fungicide mainly used against rice blast disease. Its mode of action involves inhibiting respiratory electron transfer.

Agriculture

Specifications

Product Line PESTANAL
CAS 133408-50-1
Physical Form Neat
Linear Formula C16H16N2O3
MDL Number MFCD03792728
Synonym Methyl (E)-alpha-methoxyimino-N-methyl-2-phenoxyphenylacetamide
Type Analytical Standard
Quantity 10 mg
Formula Weight 284.31
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH