missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Diuron-d6, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 1007536-67-5 |
| Physical Form | Neat |
| Linear Formula | C9H4D6 Cl2N2 O |
| MDL Number | MFCD01632291 |
| Synonym | 3-(3,4-Dichlorophenyl)-1,1-dimethylurea-d6 |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 239.13 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH