missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Dithianon, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 3347-22-6 |
| Physical Form | Neat |
| Linear Formula | C14H4N2O2S2 |
| MDL Number | MFCD00055492 |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 296.32 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH