missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Disulfoton-sulfone, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 2497-06-5 |
| Physical Form | Neat |
| Linear Formula | C8H19O4 PS3 |
| MDL Number | MFCD00155401 |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 306.40 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH