missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Diquat dibromide monohydrate, MilliporeSigma™ Supelco™

Catalog No. 111019597
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019597 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019597 Supplier Merck Emd Millipore Supplier No. 45422250MGR
Agriculture

Specifications

Product Line PESTANAL
CAS 6385-62-2
Physical Form Neat
Linear Formula C12H12 Br2N2 · H2O
MDL Number MFCD00211319
Synonym Diquat
Type Analytical Standard
Quantity 250 mg
Formula Weight 362.06
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH