missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Dinoseb, MilliporeSigma™ Supelco™

Catalog No. 111020361
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020361 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020361 Supplier Merck Emd Millipore Supplier No. 45453100MG
Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Product Line PESTANAL
CAS 88-85-7
Flash Point 152°C
Physical Form Neat
Linear Formula C10H12N2O5
MDL Number MFCD00024246
Synonym 2-sec-Butyl-4,6-dinitrophenol
Type Analytical Standard
Quantity 100 mg
Formula Weight 240.21
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH