missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Dimethylvinphos, MilliporeSigma™ Supelco™
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 2274-67-1 |
| Physical Form | Neat |
| Linear Formula | C10H10 Cl3O4 P |
| MDL Number | MFCD00272599 |
| Synonym | 2-Chloro-1-(2,4-dichlorophenyl)vinyl dimethyl phosphate; Dimethylvinfos |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 331.52 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH