missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Dimethylpropiothetin hydrochloride, ≥96% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014199
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014199 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014199 Supplier Merck Emd Millipore Supplier No. 80828100MG
Only null left

Dimethylpropiothetin is a natural product found in the organisms of saline environment such as algae and aquatic plants.

Specifications

CAS 4337-33-1
Product Line PESTANAL
Linear Formula C5H10O2 S· HCl
MDL Number MFCD00142888
Synonym DMSP; Dimethyl-3-propiothetin chloride; Dimethylsulfoniopropionate hydrochloride
Type Analytical Standard
Quantity 100 mg
Formula Weight 170.66
Percent Purity ≥96% (HPLC)
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH