missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Dimethomorph, MilliporeSigma™ Supelco™

Catalog No. 111019316
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019316 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019316 Supplier Merck Emd Millipore Supplier No. 46027100MG

Dimethomorph, a cinnamic acid derivative, is a systemic morpholine fungicide mainly used against peronosporomycetes plant pathogens.

Agriculture

Specifications

Product Line PESTANAL
CAS 110488-70-5
Physical Form Neat
Linear Formula C21H22 ClNO4
MDL Number MFCD01632781
Type Analytical Standard
Quantity 100 mg
Formula Weight 387.86
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH