missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Dimethenamid, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 87674-68-8 |
| Physical Form | Neat |
| Linear Formula | C12H18 ClNO2 S |
| MDL Number | MFCD01632376 |
| Synonym | 2-Chloro-N-(2,4-dimethyl-3-thienyl)-N-(2-methoxy-1-methylethyl)acetamide |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 275.79 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH