missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Dicyclohexyl phthalate-3,4,5,6-d4, MilliporeSigma™ Supelco™
Description
Agriculture and Environmental
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 358731-25-6 |
| Physical Form | Neat |
| Linear Formula | C20D4H22O4 |
| MDL Number | MFCD01861635 |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 334.44 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH