missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Dichlorvos-(dimethyl-d6), ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018156
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018156 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018156 Supplier Merck Emd Millipore Supplier No. 7486710MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Agriculture

Specifications

Product Line PESTANAL
CAS 203645-53-8
Flash Point 100°C - Closed Cup
Linear Formula C4D6 HCl2O4 P
MDL Number MFCD00209708
Synonym 2,2-Dichlorovinyl di(methyl-d3) phosphate; Dichlorvos-d6
Type Analytical Standard
Quantity 10 mg
Formula Weight 227.01
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH