missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Dichlorprop-methyl ester, MilliporeSigma™ Supelco™

Catalog No. 111013645
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013645 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013645 Supplier Merck Emd Millipore Supplier No. 45437250MG
Agriculture

Specifications

Product Line PESTANAL
CAS 23844-57-7
Physical Form Neat
Linear Formula C10H10 Cl2O3
MDL Number MFCD00055254
Synonym Methyl 2-(2,4-dichlorophenoxy)propionate
Type Analytical Standard
Quantity 250 mg
Formula Weight 249.09
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH