missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Dichloroprop-P, MilliporeSigma™ Supelco™

Catalog No. 111013204
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013204 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013204 Supplier Merck Emd Millipore Supplier No. 31237250MG
Only null left

Dichloroprop-P is a selective, systemic and post-emergence herbicide, used for weed control management.

Agriculture

Specifications

Product Line PESTANAL
CAS 15165-67-0
Physical Form Neat
Linear Formula C9H8 Cl2O3
MDL Number MFCD00144036
Synonym (R)-(+)-2-(2,4-Dichlorophenoxy)propionic acid
Type Analytical Standard
Quantity 250 mg
Formula Weight 235.06
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH