missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Dicamba methyl ester, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 6597-78-0 |
| Physical Form | Neat |
| Linear Formula | C9H8 Cl2O3 |
| MDL Number | MFCD00056244 |
| Synonym | Methyl 3,6-dichloro-2-methoxybenzoate |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 235.06 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH