missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Diazinon-(diethyl-d10), ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018019
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018019 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018019 Supplier Merck Emd Millipore Supplier No. 743325MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Isotope labeled analytical standard for pesticide residue analysis.

Agriculture

Specifications

Chemical Name or Material Diazinon-(diethyl-d10)
CAS 100155-47-3
Product Line PESTANAL
Flash Point 112.8°C - Closed Cup
Linear Formula C12D10H11N2O3 PS
Technique Mass Spectrometry
MDL Number MFCD00144095
Boiling Point 83°C to 84°C (0.002 mmHg)
Type Analytical Standard
Industry Type Environmental Testing
Molecular Weight (g/mol) 314.41
Quantity 5 mg
Formula Weight 314.41
Percent Purity ≥98.0% (GC)
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH