missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Daminozide-(dimethyl-d6) hydrochloride, Analytical Standard, ≥97.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111003677
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111003677 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111003677 Supplier Merck Emd Millipore Supplier No. 767345MG

Specifications

Product Line PESTANAL
Linear Formula C6D6H6N2O3 · HCl
Synonym Succinic acid mono[2,2-di(methyl-d3)hydrazide] hydrochloride
Quantity 5 mg
Formula Weight 202.67
Percent Purity ≥97.0% (GC)
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH