missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Dalapon-methyl, MilliporeSigma™ Supelco™

Catalog No. 111020133
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020133 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020133 Supplier Merck Emd Millipore Supplier No. 45383250MG
Only null left
Agriculture

Specifications

Product Line PESTANAL
CAS 17640-02-7
Flash Point 40°C
Physical Form Neat
Linear Formula CH3 CCl2 COOCH3
MDL Number MFCD00144041
Synonym Methyl 2,2-dichloropropionate; Dalapon-methyl
Type Analytical Standard
Quantity 250 mg
Formula Weight 157.00
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH