missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Cypermethrin, MilliporeSigma™ Supelco™

Catalog No. 111019603
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019603 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019603 Supplier Merck Emd Millipore Supplier No. 36128100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cypermethrin is a synthetic pyrethroid pesticide, which can find applications in pest control for agriculture in the protection of cotton and vegetable crops. It can also be used in sanitary pest control.

Agriculture and Environmental

Specifications

Product Line PESTANAL
CAS 52315-07-8
Physical Form Neat
Linear Formula C22H19 Cl2 NO3
MDL Number MFCD00055328
Synonym Alphamethrine
Type Analytical Standard
Quantity 100 mg
Formula Weight 416.30
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH