missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Cymoxanil, MilliporeSigma™ Supelco™

Catalog No. 111019098
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019098 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019098 Supplier Merck Emd Millipore Supplier No. 34326100MG
Only null left

Cymoxanil is an agricultural fungicide effective against Peronosporales fungus affecting fruits and vegetables.

Agriculture

Specifications

Product Line PESTANAL
CAS 57966-95-7
Physical Form Neat
Linear Formula C7H10N4O3
MDL Number MFCD00137381
Synonym 1-(2-Cyano-2-methoxyiminoacetyl)-3-ethylurea
Type Analytical Standard
Quantity 100 mg
Formula Weight 198.18
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH