missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Cycloprothrin, MilliporeSigma™ Supelco™

Catalog No. 111018468
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018468 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018468 Supplier Merck Emd Millipore Supplier No. 3706010MG
Only null left

Cycloprothrin is an insecticide, used for controlling insect pests on rice plants and vegetables.

Specifications

Product Line PESTANAL
CAS 63935-38-6
Physical Form Neat
Linear Formula C26H21 Cl2 NO4
MDL Number MFCD00210269
Synonym (RS)-alpha-Cyano-3-phenoxybenzyl (RS)-2,2-dichloro-1-(4-ethoxyphenyl)cyclopropanecarboxylate
Type Analytical Standard
Quantity 10 mg
Formula Weight 482.36
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH