missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Coumatetralyl, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 5836-29-3 |
| Flash Point | >100°C |
| Physical Form | Neat |
| Linear Formula | C19H16O3 |
| MDL Number | MFCD09954502 |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 292.33 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH