missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Cloxyfonac, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 6386-63-6 |
| Physical Form | Neat |
| Linear Formula | C9H9 ClO4 |
| MDL Number | MFCD01671830 |
| Synonym | 2-[4-Chloro-2-(hydroxymethyl)phenoxy]acetic acid; 4-Chloro-alpha-hydroxy-o-tolyloxyacetic acid |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 216.62 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH