missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Clothianidin, MilliporeSigma™ Supelco™

Catalog No. 111018644
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018644 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018644 Supplier Merck Emd Millipore Supplier No. 33589100MG
Only null left

Find more information here: Neonicotinoids

Agriculture

Specifications

Product Line PESTANAL
CAS 210880-92-5
Physical Form Neat
Linear Formula C6H8 ClN5O2 S
MDL Number MFCD06200753
Synonym (E)-1-(2-Chloro-5-thiazolylmethyl)-3-methyl-2-nitroguanidine
Type Analytical Standard
Quantity 100 mg
Formula Weight 249.68
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH