missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Clothianidin-d3, Analytical Standard, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018424
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018424 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018424 Supplier Merck Emd Millipore Supplier No. 5681650MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Find more information here: Neonicotinoids

Specifications

Packaging Bottomless Glass Bottle with Fused Cone
CAS 1262776-24-8
Product Line PESTANAL
Physical Form Neat
Linear Formula C6D3H5ClN5O2S
MDL Number MFCD17019117
Synonym (E)-1-(2-Chloro-5-thiazolylmethyl)-3-(methyl-d3)-2-nitroguanidine
Quantity 50 mg
Formula Weight 252.70
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH