missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Closantel, MilliporeSigma™ Supelco™
Description
Agriculture
Spécifications
Spécifications
| Product Line | PESTANAL |
| CAS | 57808-65-8 |
| Physical Form | Neat |
| Linear Formula | C22H14 Cl2I2N2O2 |
| MDL Number | MFCD00661151 |
| Synonym | 5 ′-Chloro-4 ′-(4-chloro-alpha-cyanobenzyl)-3,5-diiodo-2 ′-methylsalicylanilide; N-[5-Chloro-4-(4-chloro-alpha-cyanobenzyl)-2-methylphenyl]-2-hydroxy-3,5-diiodobenzamide |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 663.07 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH