missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Clodinafop-propargyl, MilliporeSigma™ Supelco™

Catalog No. 111019341
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019341 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019341 Supplier Merck Emd Millipore Supplier No. 31676250MG
Only null left

Clodinafop-propargyl is a foliar-absorbed aryloxyphenoxypropionate herbicide which has the capacity to inhibit the lipid biosynthesizing enzyme, acetyl coenzyme-A-carboxylase.1

Agriculture

Specifications

Product Line PESTANAL
CAS 105512-06-9
Physical Form Neat
Linear Formula C17H13 ClFNO4
MDL Number MFCD01632328
Synonym 2-Propynyl (R)-2-[4-(5-chloro-3-fluoro-2-pyridinyloxy)phenoxy]propionate
Type Analytical Standard
Quantity 250 mg
Formula Weight 349.74
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH